5-Cyano-2-fluorobenzaldehyde 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC201142-25G
APOSPC201142-25GEA 172 EUR
APOSPC201142-25G
5-Cyano-2-fluorobenzaldehyde 95+%
5-Cyano-2-fluorobenzaldehyde
Formule: C₈H₄FNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04974126
CAS nummer: 146137-79-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR