6-Chloro-3-(trifluoromethyl)picolinonitrile 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC201249-250MG PC201249-1G PC201249-5G
APOSPC201249-250MGEA 81.8 EUR
APOSPC201249-250MG APOSPC201249-1G APOSPC201249-5G
6-Chloro-3-(trifluoromethyl)picolinonitrile 95+%
6-Chloro-3-(trifluoromethyl)picolinonitrile
Formule: C₇H₂ClF₃N₂
MDL Nummer: MFCD18261725
CAS nummer: 401590-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR