2-Amino-3-(trifluoromethyl)benzonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC2036-5G PC2036-100G PC2036-25G
APOSPC2036-5GEA 67.7 EUR
APOSPC2036-5G APOSPC2036-100G APOSPC2036-25G
2-Amino-3-(trifluoromethyl)benzonitrile 95%
2-Amino-3-(trifluoromethyl)benzonitrile
Formule: C₈H₅F₃N₂
Smeltpunt: 62…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04972665
CAS nummer: 58458-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR