Copper(II) fluoride, watervrij 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC2100-100G PC2100-25G PC2100-5G
APOSPC2100-100GEA 380 EUR
APOSPC2100-100G APOSPC2100-25G APOSPC2100-5G
Copper(II) fluoride, watervrij 99%
Copper(II) fluoride
Formule: CuF₂
Dichtheid: 4,23 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010977
CAS nummer: 7789-19-7
EINECS: 232-147-3
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR