2-Fluoro-4,6-diiodoaniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC210036-1G PC210036-500MG
APOSPC210036-1GEA 1070 EUR
APOSPC210036-1G APOSPC210036-500MG
2-Fluoro-4,6-diiodoaniline
2-Fluoro-4,6-diiodoaniline
Formule: C₆H₄FI₂N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD14708138
CAS nummer: 1301739-25-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR