3-(Trifluoromethoxy)benzyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC2637-1G PC2637-5G PC2637-25G
APOSPC2637-1GEA 129 EUR
APOSPC2637-1G APOSPC2637-5G APOSPC2637-25G
3-(Trifluoromethoxy)benzyl chloride
3-(Trifluoromethoxy)benzyl chloride
Formule: C₈H₆ClF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08458087
CAS nummer: 89807-43-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR