4-Bromo-2-fluoro-5-nitroaniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC2673-1G PC2673-5G PC2673-25G
APOSPC2673-1GEA 27.8 EUR
APOSPC2673-1G APOSPC2673-5G APOSPC2673-25G
4-Bromo-2-fluoro-5-nitroaniline
4-Bromo-2-fluoro-5-nitroaniline
Formule: C₆H₄BrFN₂O₂
MW: 235.0106 g/mol
Smeltpunt: 92…94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08741367
CAS nummer: 87547-06-6
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR