2,3'-Difluorobenzophenone 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC2689M-1G PC2689M-5G
APOSPC2689M-1GEA 99.3 EUR
APOSPC2689M-1G APOSPC2689M-5G
2,3'-Difluorobenzophenone 97%
2,3'-Difluorobenzophenone
Formule: C₁₃H₈F₂O
Kookpunt: 142…143 °C/3mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00017914
CAS nummer: 58139-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR