2-Chloro-5-fluorobenzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC2717-5G PC2717-1G PC2717-25G
APOSPC2717-5GEA 26.3 EUR
APOSPC2717-5G APOSPC2717-1G APOSPC2717-25G
2-Chloro-5-fluorobenzonitrile 98%
2-Chloro-5-fluorobenzonitrile
Formule: C₇H₃ClFN
Smeltpunt: 83…85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094168
CAS nummer: 57381-56-3
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR