2,5-Difluorobenzoyl chloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC2724-10G PC2724-100G PC2724-25G PC2724-5G
APOSPC2724-10GEA 38.1 EUR
APOSPC2724-10G APOSPC2724-100G APOSPC2724-25G APOSPC2724-5G
2,5-Difluorobenzoyl chloride 98%
2,5-Difluorobenzoyl chloride
Formule: C₇H₃ClF₂O
Kookpunt: 92…93 °C/34mmHg
Dichtheid: 1,425 g/cm³ (25 °C)
Vlampunt: 59 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009929
CAS nummer: 35730-09-7
EINECS: 252-701-8
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR