trans-2,5-Difluorocinnamic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC2762-25G PC2762-100G PC2762-5G
APOSPC2762-25GEA 77.5 EUR
APOSPC2762-25G APOSPC2762-100G APOSPC2762-5G
trans-2,5-Difluorocinnamic acid 97%
trans-2,5-Difluorocinnamic acid
Formule: C₉H₆F₂O₂
Smeltpunt: 138…140 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010318
CAS nummer: 112898-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR