3-Bromo-5-nitrobenzyl fluoride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC27896-1G PC27896-250MG
APOSPC27896-1GEA 614 EUR
APOSPC27896-1G APOSPC27896-250MG
3-Bromo-5-nitrobenzyl fluoride 95%
3-Bromo-5-nitrobenzyl fluoride
Formule: C₇H₅BrFNO₂
MW: 234.023 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD31664893
CAS nummer: 2244084-96-4
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR