(2Z)-2-{[2-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC27956-1G
APOSPC27956-1GEA 1580 EUR
APOSPC27956-1G
(2Z)-2-{[2-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech
(2Z)-2-{[2-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol
Formule: C₁₅H₁₆F₃NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02081948
CAS nummer: 477848-11-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR