Methyl amino(3-fluorophenyl)acetate hydrochloride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC28213-1G PC28213-5G
APOSPC28213-1GEA 338 EUR
APOSPC28213-1G APOSPC28213-5G
Methyl amino(3-fluorophenyl)acetate hydrochloride 95%
Methyl amino(3-fluorophenyl)acetate hydrochloride
Formule: C₉H₁₁ClFNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09152736
CAS nummer: 42718-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR