Potassium(4-chlorophenyl)trifluoroborate 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC2917-10G PC2917-25G PC2917-5G
APOSPC2917-10GEA 150 EUR
APOSPC2917-10G APOSPC2917-25G APOSPC2917-5G
Potassium(4-chlorophenyl)trifluoroborate 95+%
Potassium(4-chlorophenyl)trifluoroborate
Formule: C₆H₄BClF₃K
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115770
CAS nummer: 661465-44-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR