2-Bromo-6-fluoroanisole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Bromo-3-fluoro-2-methoxybenzene

PC2958-25G
APOSPC2958-25GEA 129 EUR
APOSPC2958-25G
2-Bromo-6-fluoroanisole
2-Bromo-6-fluoroanisole
Formule: C₇H₆BrFO
MW: 205.03 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04974125
CAS nummer: 845829-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR