1,1,2,2-Tetrafluoro-1,2-diiodoethane 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC2980-25G PC2980-5G
APOSPC2980-25GEA 503 EUR
APOSPC2980-25G APOSPC2980-5G
1,1,2,2-Tetrafluoro-1,2-diiodoethane 97%
1,1,2,2-Tetrafluoro-1,2-diiodoethane
Formule: C₂F₄I₂
Kookpunt: 112…113 °C
Smeltpunt: –21 °C
Dichtheid: 2,629 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00039401
CAS nummer: 354-65-4
EINECS: 205-567-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR