4-Fluoro-o-toluic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC2983-100G PC2983-25G PC2983-5G
APOSPC2983-100GEA 109 EUR
APOSPC2983-100G APOSPC2983-25G APOSPC2983-5G
4-Fluoro-o-toluic acid 97%
4-Fluoro-o-toluic acid
Formule: C₈H₇FO₂
Smeltpunt: 168…172 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115880
CAS nummer: 321-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR