1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole-2-sulphonyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC300855-1G
APOSPC300855-1GEA 436 EUR
APOSPC300855-1G
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole-2-sulphonyl chloride
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole-2-sulphonyl chloride
Formule: C₁₁H₅Cl₃F₃NO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28954647
CAS nummer: 1858251-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR