1-[2-Methyl-3-(trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC300919-1G
APOSPC300919-1GEA 500 EUR
APOSPC300919-1G
1-[2-Methyl-3-(trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
1-[2-Methyl-3-(trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
Formule: C₁₃H₁₀F₃NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD30481160
CAS nummer: 2197063-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR