2,4-Difluoro-5-iodobenzhydrazide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC300985-1G PC300985-250MG
APOSPC300985-1GEA 38 EUR
APOSPC300985-1G APOSPC300985-250MG
2,4-Difluoro-5-iodobenzhydrazide
2,4-Difluoro-5-iodobenzhydrazide
Formule: C₇H₅F₂IN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD31630104
CAS nummer: 2244085-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR