Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC300997-1G PC300997-250MG
APOSPC300997-1GEA 342 EUR
APOSPC300997-1G APOSPC300997-250MG
Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Formule: C₁₃H₁₃BrFNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16630891
CAS nummer: 1239731-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR