3-Fluoro-5-(trifluoromethyl)-DL-phenylalanine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302143-1G PC302143-5G PC302143-10G
APOSPC302143-1GEA 129 EUR
APOSPC302143-1G APOSPC302143-5G APOSPC302143-10G
3-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
3-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
Formule: C₁₀H₉F₄NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115997
CAS nummer: 1259992-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR