3-Chloro-5-(trifluoromethyl)benzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC302290-25G PC302290-5G PC302290-1G
APOSPC302290-25GEA 204 EUR
APOSPC302290-25G APOSPC302290-5G APOSPC302290-1G
3-Chloro-5-(trifluoromethyl)benzaldehyde 97%
3-Chloro-5-(trifluoromethyl)benzaldehyde
Formule: C₈H₄ClF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115845
CAS nummer: 477535-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR