4-Ethoxy-3-(trifluoromethyl)aniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302332-5G PC302332-10G PC302332-1G
APOSPC302332-5GEA 43.8 EUR
APOSPC302332-5G APOSPC302332-10G APOSPC302332-1G
4-Ethoxy-3-(trifluoromethyl)aniline 98%
4-Ethoxy-3-(trifluoromethyl)aniline
Formule: C₉H₁₀F₃NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08056137
CAS nummer: 2713-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR