2-Fluoro-4-(pentafluorosulphur)benzenesulphonyl chloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302336-1G PC302336-250MG
APOSPC302336-1GEA 1930 EUR
APOSPC302336-1G APOSPC302336-250MG
2-Fluoro-4-(pentafluorosulphur)benzenesulphonyl chloride 97%
2-Fluoro-4-(pentafluorosulphur)benzenesulphonyl chloride
Formule: C₆H₃ClF₆O₂S₂
MW: 320.66 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28054239
CAS nummer: 1706462-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR