2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid

PC302387-1G PC302387-5G
APOSPC302387-1GEA 106 EUR
APOSPC302387-1G APOSPC302387-5G
2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid
Formule: C₁₀H₈F₄O₃
MW: 252.16 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22201046
CAS nummer: 1373920-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR