4-Chloro-3-fluoro-DL-phenylalanine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC302580-1G PC302580-5G
APOSPC302580-1GEA 146 EUR
APOSPC302580-1G APOSPC302580-5G
4-Chloro-3-fluoro-DL-phenylalanine
4-Chloro-3-fluoro-DL-phenylalanine
Formule: C₉H₉ClFNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06660180
CAS nummer: 439587-16-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR