4-Methoxy-2-(trifluoromethoxy)benzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302598-5G PC302598-1G PC302598-25G
APOSPC302598-5GEA 161 EUR
APOSPC302598-5G APOSPC302598-1G APOSPC302598-25G
4-Methoxy-2-(trifluoromethoxy)benzonitrile 98%
4-Methoxy-2-(trifluoromethoxy)benzonitrile
Formule: C₉H₆F₃NO₂
Smeltpunt: 31…33 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06660284
CAS nummer: 886502-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR