5-(Difluoromethoxy)-2-fluorobenzyl bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC302610-1G PC302610-5G
APOSPC302610-1GEA 261 EUR
APOSPC302610-1G APOSPC302610-5G
5-(Difluoromethoxy)-2-fluorobenzyl bromide
5-(Difluoromethoxy)-2-fluorobenzyl bromide
Formule: C₈H₆BrF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09832361
CAS nummer: 1017779-36-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR