2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302692-10G PC302692-5G PC302692-1G PC302692-25G PC302692-100G
APOSPC302692-10GEA 471 EUR
APOSPC302692-10G APOSPC302692-5G APOSPC302692-1G APOSPC302692-25G APOSPC302692-100G
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid 98%
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid
Formule: C₁₀H₆F₆O₃
Smeltpunt: 156…159 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00728738
CAS nummer: 180134-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR