4-Ethoxy-3,5-difluorobenzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC302749-10G PC302749-5G PC302749-1G PC302749-25G PC302749-100G
APOSPC302749-10GEA 284 EUR
APOSPC302749-10G APOSPC302749-5G APOSPC302749-1G APOSPC302749-25G APOSPC302749-100G
4-Ethoxy-3,5-difluorobenzoic acid 97%
4-Ethoxy-3,5-difluorobenzoic acid
Formule: C₉H₈F₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09258683
CAS nummer: 1017779-19-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR