4-Methoxy-2-(trifluoromethyl)benzyl alcohol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC302755-10G PC302755-5G PC302755-1G
APOSPC302755-10GEA 250 EUR
APOSPC302755-10G APOSPC302755-5G APOSPC302755-1G
4-Methoxy-2-(trifluoromethyl)benzyl alcohol 98%
4-Methoxy-2-(trifluoromethyl)benzyl alcohol
Formule: C₉H₉F₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06202983
CAS nummer: 773871-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR