3-Methyl-5-(trifluoromethyl)benzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC302853-10G PC302853-5G PC302853-1G
APOSPC302853-10GEA 337 EUR
APOSPC302853-10G APOSPC302853-5G APOSPC302853-1G
3-Methyl-5-(trifluoromethyl)benzaldehyde 97%
3-Methyl-5-(trifluoromethyl)benzaldehyde
Formule: C₉H₇F₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16652409
CAS nummer: 116070-39-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR