2-Methoxy-6-(trifluoromethyl)benzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC302931-10G PC302931-5G PC302931-25G PC302931-100G
APOSPC302931-10GEA 385 EUR
APOSPC302931-10G APOSPC302931-5G APOSPC302931-25G APOSPC302931-100G
2-Methoxy-6-(trifluoromethyl)benzaldehyde
2-Methoxy-6-(trifluoromethyl)benzaldehyde
Formule: C₉H₇F₃O₂
Smeltpunt: 46…49 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09832324
CAS nummer: 1017778-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR