3-Ethoxy-2,4-difluorobenzyl bromide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302952-10G PC302952-5G PC302952-1G
APOSPC302952-10GEA 643 EUR
APOSPC302952-10G APOSPC302952-5G APOSPC302952-1G
3-Ethoxy-2,4-difluorobenzyl bromide 97%
3-Ethoxy-2,4-difluorobenzyl bromide
Formule: C₉H₉BrF₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09258729
CAS nummer: 1017778-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR