3-Ethoxy-2,6-difluorobenzyl bromide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC302953-10G PC302953-5G PC302953-1G
APOSPC302953-10GEA 318 EUR
APOSPC302953-10G APOSPC302953-5G APOSPC302953-1G
3-Ethoxy-2,6-difluorobenzyl bromide 98%
3-Ethoxy-2,6-difluorobenzyl bromide
Formule: C₉H₉BrF₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11519372
CAS nummer: 1092461-30-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR