2,4-Difluoro-3-methoxybenzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303018-10G PC303018-5G PC303018-1G
APOSPC303018-10GEA 1060 EUR
APOSPC303018-10G APOSPC303018-5G APOSPC303018-1G
2,4-Difluoro-3-methoxybenzonitrile 98%
2,4-Difluoro-3-methoxybenzonitrile
Formule: C₈H₅F₂NO
Smeltpunt: 44…46 °C
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 220353-20-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR