4-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303091-1G
APOSPC303091-1GEA 424 EUR
APOSPC303091-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
Formule: C₉H₅F₄N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22201068
CAS nummer: 1373920-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR