4-Methoxy-3-methyl-5-(trifluoromethyl)benzoyl chloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303093-5G PC303093-1G
APOSPC303093-5GEA 309 EUR
APOSPC303093-5G APOSPC303093-1G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzoyl chloride 98%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzoyl chloride
Formule: C₁₀H₈ClF₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22682812
CAS nummer: 1431329-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR