6-Chloro-2,3-difluorobenzyl bromide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303099-10G PC303099-5G PC303099-1G
APOSPC303099-10GEA 353 EUR
APOSPC303099-10G APOSPC303099-5G APOSPC303099-1G
6-Chloro-2,3-difluorobenzyl bromide 97%
6-Chloro-2,3-difluorobenzyl bromide
Formule: C₇H₄BrClF₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22201040
CAS nummer: 1373921-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR