4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303109-10G PC303109-5G PC303109-1G
APOSPC303109-10GEA 408 EUR
APOSPC303109-10G APOSPC303109-5G APOSPC303109-1G
4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline 97%
4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline
Formule: C₁₃H₉F₄NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08687913
CAS nummer: 946784-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR