2-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-Chloro-3-(chlorosulphonyl)benzotrifluoride

PC303179-1G PC303179-250MG
APOSPC303179-1GEA 443 EUR
APOSPC303179-1G APOSPC303179-250MG
2-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride
Formule: C₇H₃Cl₂F₃O₂S
MW: 279.064 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13185469
CAS nummer: 1214346-10-7
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR