2-Methoxy-5-(trifluoromethyl)benzenesulphonyl chloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC303181-1G PC303181-250MG
APOSPC303181-1GEA 680 EUR
APOSPC303181-1G APOSPC303181-250MG
2-Methoxy-5-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Methoxy-5-(trifluoromethyl)benzenesulphonyl chloride
Formule: C₈H₆ClF₃O₃S
MW: 274.645 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06637310
CAS nummer: 612541-12-5
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR