2-Chloro-4-fluoro-5-methylbenzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303345-1G PC303345-5G
APOSPC303345-1GEA 275 EUR
APOSPC303345-1G APOSPC303345-5G
2-Chloro-4-fluoro-5-methylbenzonitrile 97%
2-Chloro-4-fluoro-5-methylbenzonitrile
Formule: C₈H₅ClFN
MW: 169.583 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD21602109
CAS nummer: 924626-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR