4'-Chloro-2'-(trifluoromethyl)acetophenone 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303395-10G PC303395-1G PC303395-5G PC303395-25G
APOSPC303395-10GEA 363 EUR
APOSPC303395-10G APOSPC303395-1G APOSPC303395-5G APOSPC303395-25G
4'-Chloro-2'-(trifluoromethyl)acetophenone 97%
4'-Chloro-2'-(trifluoromethyl)acetophenone
Formule: C₉H₆ClF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13194258
CAS nummer: 86520-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR