4'-Chloro-3'-(trifluoromethyl)propiophenone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303396-10G PC303396-1G PC303396-5G PC303396-25G
APOSPC303396-10GEA 1460 EUR
APOSPC303396-10G APOSPC303396-1G APOSPC303396-5G APOSPC303396-25G
4'-Chloro-3'-(trifluoromethyl)propiophenone 98%
4'-Chloro-3'-(trifluoromethyl)propiophenone
Formule: C₁₀H₈ClF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18393944
CAS nummer: 1261861-70-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR