5-Bromo-3-chloro-2-fluorobenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (5-Bromo-3-chloro-2-fluorophenyl)methanol

PC303453-1G PC303453-5G
APOSPC303453-1GEA 233 EUR
APOSPC303453-1G APOSPC303453-5G
5-Bromo-3-chloro-2-fluorobenzyl alcohol
5-Bromo-3-chloro-2-fluorobenzyl alcohol
Formule: C₇H₅BrClFO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD29036608
CAS nummer: 1514304-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR