5-Bromo-3-chloro-2-fluorophenylacetonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303455-1G PC303455-5G
APOSPC303455-1GEA 328 EUR
APOSPC303455-1G APOSPC303455-5G
5-Bromo-3-chloro-2-fluorophenylacetonitrile
5-Bromo-3-chloro-2-fluorophenylacetonitrile
Formule: C₈H₄BrClFN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD26686147
CAS nummer: 1537627-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR