3-Bromo-4-(trifluoromethoxy)phenylacetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC303526-1G PC303526-5G
APOSPC303526-1GEA 421 EUR
APOSPC303526-1G APOSPC303526-5G
3-Bromo-4-(trifluoromethoxy)phenylacetic acid
3-Bromo-4-(trifluoromethoxy)phenylacetic acid
Formule: C₉H₆BrF₃O₃
MW: 299.041 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD32640115
CAS nummer: 2169367-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR